1-[(3-Fluorophenyl)methyl]-4-(furan-2-yl)-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSPC28194-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:294.28 g/mol
Storage Temperature:Ambient
CAS Number:439120-70-4
Formula:C₁₇H₁₁FN₂O₂
MDL Number:MFCD03012366