Order Entry
Poland
ContactUsLinkComponent
1-[(4-Fluorophenyl)methyl]-4-(furan-2-yl)-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
1-[(4-Fluorophenyl)methyl]-4-(furan-2-yl)-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSPC28179-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[(4-Fluorophenyl)methyl]-4-(furan-2-yl)-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSPC28179-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :294.28 g/mol
Storage Temperature:Ambient
CAS Number:478259-93-7
Formula:C₁₇H₁₁FN₂O₂
MDL Number:MFCD02571450