Order Entry
Poland
ContactUsLinkComponent
4-{[2-Oxo-5-(trifluoromethyl)-1,2-dihydropyridin-1-yl]methyl}benzoic acid, tech
4-{[2-Oxo-5-(trifluoromethyl)-1,2-dihydropyridin-1-yl]methyl}benzoic acid, tech
Catalog # APOSPC28167-1G
Supplier:  Apollo Scientific
CAS Number:  
4-{[2-Oxo-5-(trifluoromethyl)-1,2-dihydropyridin-1-yl]methyl}benzoic acid, tech
Catalog # APOSPC28167-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :297.229 g/mol
Storage Temperature:Ambient
CAS Number:339024-98-5
Formula:C₁₄H₁₀F₃NO₃
MDL Number:MFCD00140124