Order Entry
Poland
ContactUsLinkComponent
Methyl 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetate, tech
Methyl 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetate, tech
Catalog # APOSPC28032-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetate, tech
Catalog # APOSPC28032-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :258.197 g/mol
Storage Temperature:Ambient
CAS Number:139591-08-5
Formula:C₁₁H₉F₃N₂O₂
MDL Number:MFCD02102654