(2Z)-2-[(2-Fluorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-ol, tech
Catalog # APOSPC27958-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.281 g/mol
Storage Temperature:Ambient
CAS Number:477848-27-4
Formula:C₁₄H₁₆FNO
MDL Number:MFCD02081952