1-([(9H-Fluoren-9-ylmethoxy)carbonyl]amino)-3,3-difluorocyclobutane-1-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1936532-04-5
:
Formula:C₂₀H₁₇F₂NO₄
MDL Number:MFCD28389003
:373.35 g/mol
Storage Temperature:Ambient
Frequently Bought Together