
2-(Bicyclo[2.2.1]hept-5-en-2-yl)-1,1,1-trifluoro-2-(trifluoromethyl)propan-2-ol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:196314-61-1
:
Formula:C₁₁H₁₂F₆O
MDL Number:MFCD07777113
:274.203 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together