Order Entry
Poland
ContactUsLinkComponent
2,4-Dichloro-5-(difluoromethyl)pyridine
Irritant
2,4-Dichloro-5-(difluoromethyl)pyridine
Catalog # APOSPC200458-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,4-Dichloro-5-(difluoromethyl)pyridine
Catalog # APOSPC200458-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :197.998 g/mol
Melting Pt:37…40 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1256833-30-3
Formula:C₆H₃Cl₂F₂N
MDL Number:MFCD18257012