Order Entry
Poland
ContactUsLinkComponent
1-[5-Chloro-4-(difluoromethyl)pyridin-2-yl]piperazine
1-[5-Chloro-4-(difluoromethyl)pyridin-2-yl]piperazine
Catalog # APOSPC200448-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[5-Chloro-4-(difluoromethyl)pyridin-2-yl]piperazine
Catalog # APOSPC200448-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :247.672 g/mol
Storage Temperature:Ambient
CAS Number:1565827-81-7
Formula:C₁₀H₁₂ClF₂N₃
MDL Number:MFCD27957335