Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dibromo-2-fluoro-4-methoxybenzene
1,3-Dibromo-2-fluoro-4-methoxybenzene
1,3-Dibromo-2-fluoro-4-methoxybenzene
APOSPC200430-500MG
:
Apollo Scientific
:
1,3-Dibromo-2-fluoro-4-methoxybenzene
APOSPC200430-500MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
500 mg
Seq:
2725
:
:
283.92 g/mol
:
92…94 °C
:
Ambient
:
1526600-49-6
:
C₇H₅Br₂FO
:
MFCD27957293