Potassium (2,6-difluorophenyl)trifluoroboranuide
Catalog # APOSPC200387-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Rodzaj opakowania:Bottle
- Ilość:1 g
- Seq:3047
Specifications
About this item
:
:219.989 g/mol
Storage Temperature:Temperatura otoczenia
CAS Number:267006-25-7
Formula:C₆H₃BF₅·K
MDL Number:MFCD05865179