Order Entry
Poland
ContactUsLinkComponent
(S)-N-Methyl-1-[3,5-bis(trifluoromethyl)phenyl]ethylamine
Irritant
(S)-N-Methyl-1-[3,5-bis(trifluoromethyl)phenyl]ethylamine
Catalog # APOSPC1166-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(S)-N-Methyl-1-[3,5-bis(trifluoromethyl)phenyl]ethylamine
Catalog # APOSPC1166-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :271.202 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:511256-36-3
Formula:C₁₁H₁₁F₆N
MDL Number:MFCD07779108