3-[4-(2,3,4-Trifluorophenoxy)phenyl]-1H-pyrazole
Catalog # APOSPC110033-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Rodzaj opakowania:Bottle
- Ilość:1 g
- Seq:3047
Specifications
About this item
:
:290.24 g/mol
Storage Temperature:Temperatura otoczenia
Formula:C₁₅H₉F₃N₂O
MDL Number:MFCD32649084