
2-Bromo-1,4-difluoro-3-(trifluoromethoxy)benzene 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1807078-27-8
:
Formula:C₇H₂BrF₅O
MDL Number:MFCD28738359
:276.986 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together