Order Entry
Poland
ContactUsLinkComponent
 
2-Bromo-6-fluorophenylhydrazine hydrochloride 97%
Catalog # APOSPC10332-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-6-fluorophenylhydrazine hydrochloride 97%
Catalog # APOSPC10332-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :241.489 g/mol
Storage Temperature:Ambient
CAS Number:1049729-31-8
Formula:C₆H₆BrFN₂·ClH
MDL Number:MFCD07368676