Order Entry
Poland
ContactUsLinkComponent
1,1-Dichloro-2,3,3,3-tetrafluoroprop-1-ene
1,1-Dichloro-2,3,3,3-tetrafluoroprop-1-ene
Catalog # APOSPC0965-5G
Supplier:  Apollo Scientific
CAS Number:  
1,1-Dichloro-2,3,3,3-tetrafluoroprop-1-ene
Catalog # APOSPC0965-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :182.932 g/mol
Storage Temperature:Ambient
CAS Number:2804-55-9
Formula:C₃Cl₂F₄
MDL Number:MFCD09763641