
2-Chloro-2-(trifluoromethyl)perfluoropentane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:67437-97-2
:
Formula:C₆ClF₁₃
MDL Number:MFCD00153741
Boiling Pt:83…84 °C
:354.496 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together