
(1R)-1-[3-(Trifluoromethyl)phenyl]ethylamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:127852-30-6
:
Formula:C₉H₁₀F₃N
MDL Number:MFCD03093005
:189.178 g/mol
Boiling Pt:69…71 °C (4.5 mm Hg)
Flash Pt:72 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together