
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine
Catalog # APOSPC0585-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:257.176 g/mol
Boiling Pt:63.5…64.5 °C
Density:1.1 g/cm³ (25 °C)
Flash Pt:66 °C
Storage Temperature:Refrigerator
UN:2735
ADR:8,III
:GHS05
CAS Number:127733-40-8
Formula:C₁₀H₉F₆N
MDL Number:MFCD03093013