
3,5-Dichloro-4-fluoroaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2729-34-2
:
Formula:C₆H₄Cl₂FN
MDL Number:MFCD09835492
:180.007 g/mol
Melting Pt:102 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together