Order Entry
Poland
ContactUsLinkComponent
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Catalog # APOSPC01471-1G
Supplier:  Apollo Scientific
CAS Number:  
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Catalog # APOSPC01471-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :318.272 g/mol
Storage Temperature:Ambient
CAS Number:874839-24-4
Formula:C₁₂H₉F₃N₂O₃S
MDL Number:MFCD07774165