Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Dimethylpent-4-enenitrile
2,2-Dimethylpent-4-enenitrile 97%
2,2-Dimethylpent-4-enenitrile 97%
APOSOR960348-1G
:
Apollo Scientific
:
2,2-Dimethylpent-4-enenitrile 97%
APOSOR960348-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
109.169 g/mol
:
Ambient
:
2978-30-5
:
C₇H₁₁N
:
MFCD20483740