Ethyl 2-(3-hydroxyphenoxy)acetate 95%
Catalog # APOSOR957645-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.2 g/mol
Storage Temperature:Ambient
CAS Number:68747-24-0
Formula:C₁₀H₁₂O₄
MDL Number:MFCD23111306