Order Entry
Poland
ContactUsLinkComponent
5-Chloro-3-methylthiophene-2-carbonitrile 98%
5-Chloro-3-methylthiophene-2-carbonitrile 98%
Catalog # APOSOR956957-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-3-methylthiophene-2-carbonitrile 98%
Catalog # APOSOR956957-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :157.621 g/mol
Storage Temperature:Ambient
Formula:C₆H₄ClNS
MDL Number:MFCD25972916