Order Entry
Poland
ContactUsLinkComponent
 
4-Chloro-1-cyclopropyl-1H-pyrazole 95%
Catalog # APOSOR956956-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-1-cyclopropyl-1H-pyrazole 95%
Catalog # APOSOR956956-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :142.586 g/mol
Storage Temperature:Ambient
CAS Number:1204901-66-5
Formula:C₆H₇ClN₂
MDL Number:MFCD23135674