Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-1-isopropyl-1H-pyrazole
4-Chloro-1-isopropyl-1H-pyrazole 95%
4-Chloro-1-isopropyl-1H-pyrazole 95%
4-Chloro-1-isopropyl-1H-pyrazole 95%
APOSOR956950-1G
:
Apollo Scientific
:
4-Chloro-1-isopropyl-1H-pyrazole 95%
APOSOR956950-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
144.602 g/mol
:
Ambient
:
1205921-77-2
:
C₆H₉ClN₂
:
MFCD15976410