Order Entry
Poland
ContactUsLinkComponent
Methyl 2-[[(1S)-1-phenylethyl]amino]acetate 98%
Methyl 2-[[(1S)-1-phenylethyl]amino]acetate 98%
Catalog # APOSOR955654-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-[[(1S)-1-phenylethyl]amino]acetate 98%
Catalog # APOSOR955654-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.242 g/mol
Storage Temperature:Ambient
CAS Number:248603-82-9
Formula:C₁₁H₁₅NO₂
MDL Number:MFCD18903630