Order Entry
Poland
ContactUsLinkComponent
2-(4-Methylphenyl)phenol 96%
2-(4-Methylphenyl)phenol 96%
Catalog # APOSOR954769-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Methylphenyl)phenol 96%
Catalog # APOSOR954769-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :184.234 g/mol
Storage Temperature:Ambient
CAS Number:101043-55-4
Formula:C₁₃H₁₂O
MDL Number:MFCD06802423