Order Entry
Poland
ContactUsLinkComponent
5,7-Dichloro[1,2,4]triazolo[1,5-a]pyrimidin-2-amine
Irritant
5,7-Dichloro[1,2,4]triazolo[1,5-a]pyrimidin-2-amine
Catalog # APOSOR951539-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5,7-Dichloro[1,2,4]triazolo[1,5-a]pyrimidin-2-amine
Catalog # APOSOR951539-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :203.029 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1124383-04-5
Formula:C₆H₄Cl₂N₄
MDL Number:MFCD18642394