1-(3,4-Dichlorophenyl)ethanol 95%
Catalog # APOSOR951245-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:191.055 g/mol
Storage Temperature:Ambient
CAS Number:1475-11-2
Formula:C₈H₈Cl₂O
MDL Number:MFCD00021865