tert-Butyl 2-(butan-2-ylidene)hydrazinecarboxylate 95%
Catalog # APOSOR947130-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:186.251 g/mol
Storage Temperature:Ambient
CAS Number:57699-47-5
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD14635684