Order Entry
Poland
ContactUsLinkComponent
1-(3,5-Dimethyl-1H-pyrazol-4-yl)ethanone 95%
1-(3,5-Dimethyl-1H-pyrazol-4-yl)ethanone 95%
Catalog # APOSOR943564-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(3,5-Dimethyl-1H-pyrazol-4-yl)ethanone 95%
Catalog # APOSOR943564-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :138.167 g/mol
Storage Temperature:Ambient
CAS Number:1123-48-4
Formula:C₇H₁₀N₂O
MDL Number:MFCD00234353