Order Entry
Poland
ContactUsLinkComponent
(S)-2-N-Boc-propane-1,2-diamine 97%
(S)-2-N-Boc-propane-1,2-diamine 97%
Catalog # APOSOR924219-1G
Supplier:  Apollo Scientific
CAS Number:  
(S)-2-N-Boc-propane-1,2-diamine 97%
Catalog # APOSOR924219-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :174.241 g/mol
Storage Temperature:Ambient
CAS Number:146552-71-8
Formula:C₈H₁₈N₂O₂
MDL Number:MFCD08726031