Order Entry
Poland
ContactUsLinkComponent
 
Methyl imidazo[1,5-a]pyridine-1-carboxylate 95%
  APOSOR922221-1G
 :  
Methyl imidazo[1,5-a]pyridine-1-carboxylate 95%
  APOSOR922221-1G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

 :
 :176.172 g/mol
 :Ambient
 :1039356-98-3
 :C₉H₈N₂O₂
 :MFCD15144579