1-(4-Hydroxy-phenyl)-cyclopropanecarboxylic acid 95%
Catalog # APOSOR920881-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:178.185 g/mol
Storage Temperature:Ambient
CAS Number:869970-25-2
Formula:C₁₀H₁₀O₃
MDL Number:MFCD19441896