Order Entry
Poland
ContactUsLinkComponent
4-Amino-1-methyl-1H-pyrazole-3-carbonitrile 98%
4-Amino-1-methyl-1H-pyrazole-3-carbonitrile 98%
Catalog # APOSOR920854-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Amino-1-methyl-1H-pyrazole-3-carbonitrile 98%
Catalog # APOSOR920854-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :122.128 g/mol
Storage Temperature:Ambient
CAS Number:1201935-84-3
Formula:C₅H₆N₄
MDL Number:MFCD19215684