Order Entry
Poland
ContactUsLinkComponent
2-Methylamino-1-piperidin-1-yl-ethanone hydrochloride 95%
2-Methylamino-1-piperidin-1-yl-ethanone hydrochloride 95%
Catalog # APOSOR913287-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methylamino-1-piperidin-1-yl-ethanone hydrochloride 95%
Catalog # APOSOR913287-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :192.686 g/mol
Storage Temperature:Ambient
CAS Number:1176419-66-1
Formula:C₈H₁₆N₂O·ClH
MDL Number:MFCD08060487