Order Entry
Poland
ContactUsLinkComponent
7-Methyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylic acid
Irritant
7-Methyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylic acid
Catalog # APOSOR9112-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
7-Methyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylic acid
Catalog # APOSOR9112-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :252.268 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:220465-49-6
Formula:C₁₅H₁₂N₂O₂
MDL Number:MFCD00269534