Order Entry
Poland
ContactUsLinkComponent
1-Cyclohexyl-4-nitropyrazole 97%
1-Cyclohexyl-4-nitropyrazole 97%
Catalog # APOSOR911093-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Cyclohexyl-4-nitropyrazole 97%
Catalog # APOSOR911093-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.218 g/mol
Storage Temperature:Ambient
CAS Number:97421-22-2
Formula:C₉H₁₃N₃O₂
MDL Number:MFCD24448797