Order Entry
Poland
ContactUsLinkComponent
Methyl 2-methylbenzimidazole-5-carboxylate 97%
Methyl 2-methylbenzimidazole-5-carboxylate 97%
Catalog # APOSOR909478-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-methylbenzimidazole-5-carboxylate 97%
Catalog # APOSOR909478-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :190.199 g/mol
Storage Temperature:Ambient
CAS Number:106429-51-0
Formula:C₁₀H₁₀N₂O₂
MDL Number:MFCD19685000