Order Entry
Poland
ContactUsLinkComponent
1-Cyclopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1h)-one 95%
1-Cyclopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1h)-one 95%
Catalog # APOSOR908611-250MG
Supplier:  Apollo Scientific
CAS Number:  
1-Cyclopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1h)-one 95%
Catalog # APOSOR908611-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :261.124 g/mol
Storage Temperature:Refrigerator
CAS Number:1596367-55-3
Formula:C₁₄H₂₀BNO₃
MDL Number:MFCD16996103