Order Entry
Poland
ContactUsLinkComponent
2-(2-tert-Butyldimethylsilyloxyethyl)bromobenzene 98%
2-(2-tert-Butyldimethylsilyloxyethyl)bromobenzene 98%
Catalog # APOSOR7200-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-tert-Butyldimethylsilyloxyethyl)bromobenzene 98%
Catalog # APOSOR7200-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :315.321 g/mol
Storage Temperature:Ambient
CAS Number:181021-20-5
Formula:C₁₄H₂₃BrOSi
MDL Number:MFCD09258648