
2-(Aminomethyl)-3,5-dimethylpyridine hydrochloride
Catalog # APOSOR61020-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:172.655 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1257535-52-6
Formula:C₈H₁₂N₂·ClH
MDL Number:MFCD17214191