Order Entry
Poland
ContactUsLinkComponent
Ethyl 5-chloro-1,4-dihydro-8-methyl-4-oxoquinoline-3-carboxylate
Ethyl 5-chloro-1,4-dihydro-8-methyl-4-oxoquinoline-3-carboxylate
Catalog # APOSOR59338-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 5-chloro-1,4-dihydro-8-methyl-4-oxoquinoline-3-carboxylate
Catalog # APOSOR59338-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :265.692 g/mol
Storage Temperature:Ambient
CAS Number:351893-52-2
Formula:C₁₃H₁₂ClNO₃
MDL Number:MFCD06009651