Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-1,2,6-Trimethylpiperazine dihydrochloride
cis-1,2,6-Trimethylpiperazine dihydrochloride
cis-1,2,6-Trimethylpiperazine dihydrochloride
APOSOR53062-1G
:
Apollo Scientific
:
cis-1,2,6-Trimethylpiperazine dihydrochloride
APOSOR53062-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
201.137 g/mol
:
Ambient
:
1195782-28-5
:
C₇H₁₆N₂·2ClH
:
MFCD26959087