Order Entry
Poland
ContactUsLinkComponent
 
(S)-1-(1H-Pyrazolo[3,4-b]pyridin-5-yl)ethanamine
Catalog # APOSOR52293-500MG
Supplier:  Apollo Scientific
CAS Number:  
(S)-1-(1H-Pyrazolo[3,4-b]pyridin-5-yl)ethanamine
Catalog # APOSOR52293-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :162.192 g/mol
Storage Temperature:Ambient
CAS Number:1932616-44-8
Formula:C₈H₁₀N₄
MDL Number:MFCD29042486