Order Entry
Poland
ContactUsLinkComponent
3-Iodo-6-methoxy-1H-pyrrolo[3,2-c]pyridine ≥97%
3-Iodo-6-methoxy-1H-pyrrolo[3,2-c]pyridine ≥97%
Catalog # APOSOR510051-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-Iodo-6-methoxy-1H-pyrrolo[3,2-c]pyridine ≥97%
Catalog # APOSOR510051-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :274.058 g/mol
Storage Temperature:Refrigerator
CAS Number:1190315-47-9
Formula:C₈H₇IN₂O
MDL Number:MFCD12963241