Order Entry
Poland
ContactUsLinkComponent
1-Oxa-9-azaspiro[5.5]undecane hydrochloride 95+%
Irritant
1-Oxa-9-azaspiro[5.5]undecane hydrochloride 95+%
Catalog # APOSOR500040-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Oxa-9-azaspiro[5.5]undecane hydrochloride 95+%
Catalog # APOSOR500040-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :191.698 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:71879-41-9
 :MFCD11036126
Formula:C₉H₁₇NO·ClH