
(2R)-4-Iodo-2-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine, N-BOC protected 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2366997-18-2
:
Formula:C₁₃H₁₇IN₂O₂
:360.191 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together