Order Entry
Poland
ContactUsLinkComponent
4-Bromo-6-methoxy-pyrazolo[1,5-a]pyridine
4-Bromo-6-methoxy-pyrazolo[1,5-a]pyridine
Catalog # APOSOR49064-250MG
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-6-methoxy-pyrazolo[1,5-a]pyridine
Catalog # APOSOR49064-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :227.058 g/mol
Storage Temperature:Ambient
CAS Number:1207839-86-8
Formula:C₈H₇BrN₂O
MDL Number:MFCD22689926